Merck
  • A metabonomic study of adjuvant-induced arthritis in rats using ultra-performance liquid chromatography coupled with quadrupole time-of-flight mass spectrometry.
A metabonomic study of adjuvant-induced arthritis in rats using ultra-performance liquid chromatography coupled with quadrupole time-of-flight mass spectrometry.
Molecular bioSystems (2014-07-22)
Yao Qi, Zifeng Pi, Shu Liu, Fengrui Song, Na Lin, Zhiqiang Liu
ABSTRACT
Rheumatoid arthritis (RA) is a chronic systemic inflammatory and autoimmune disease accompanied by the destruction and deformities of joints. Adjuvant-induced arthritis (AIA) is one of the excellent animal models of RA used to understand disease pathogenesis and screen potential drugs. In this paper, a urinary metabonomics method based on ultra-performance liquid chromatography coupled with quadrupole time-of-flight mass spectrometry (UPLC-Q-TOF-MS) has been established to investigate the disease progression of AIA and find potential biomarkers of secondary inflammation in AIA rats. 24 potential biomarkers were identified, including xanthurenic acid, kynurenic acid, 4-pyridoxic acid, and phenylalanine, which revealed that tryptophan metabolism, phenylalanine metabolism, gut microbiota metabolism and energy metabolism were disturbed in AIA rats. These potential biomarkers and their corresponding pathways are helpful to further understand the mechanisms of AIA and pathogenesis of RA. This study demonstrates that metabonomics based on UPLC-Q-TOF-MS is a powerful methodology to analyze the underlying disease pathogenesis.
Sigma-Aldrich
Hippuric acid, 98%
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
DL-Kynurenine, ≥95.0% (NT)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Valine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
DL-Valine, ≥97%
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
DL-Valine, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Citric acid, 99%
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Phenylalanine, European Pharmacopoeia (EP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material