Merck
Gp130-dependent signaling in the podocyte.
American journal of physiology. Renal physiology (2014-06-06)
Yoshikuni Nagayama, Gerald S Braun, Christina M Jakobs, Yuichi Maruta, Claudia R van Roeyen, Barbara M Klinkhammer, Peter Boor, Luigi Villa, Ute Raffetseder, Christian Trautwein, Dieter Görtz, Gerhard Müller-Newen, Tammo Ostendorf, Jürgen Floege
ABSTRACT
Renal inflammation, in particular glomerular, is often characterized by increased IL-6 levels. The in vivo relevance of IL-6 signaling in glomerular podocytes, which play central roles in most glomerular diseases, is unknown. Here, we show that in normal mice, podocytes express gp130, the common signal-transducing receptor subunit of the IL-6 family of cytokines. Following systemic IL-6 or LPS injection in mice, podocyte IL-6 signaling was evidenced by downstream STAT3 phosphorylation. Next, we generated mice deficient for gp130 in podocytes. Expectedly, these mice exhibited abrogated IL-6 downstream signaling in podocytes. At the age of 40 wk, they did not show spontaneous renal pathology or abnormal renal function. The mice were then challenged using two LPS injury models as well as nephrotoxic serum to induce crescentic nephritis. Under all conditions, circulating IL-6 levels increased markedly and the mice developed the pathological hallmarks of the corresponding injury models such as proteinuria and development of glomerular crescents, respectively. However, despite the capacity of normal podocytes to transduce IL-6 family signals downstream, there were no significant differences between mice bearing the podocyte-specific gp130 deletion and their control littermates in any of these models. In conclusion, under the different conditions tested, gp130 signaling was not a critical component of the (patho-)biology of the podocyte in vivo.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Anti-PCNA (Ab-1) Mouse mAb (PC10), liquid, clone PC10, Calbiochem®
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates