Merck
  • Apoptosis in ovarian granulosa cells of cattle: morphological features and clearance by homologous phagocytosis.
Apoptosis in ovarian granulosa cells of cattle: morphological features and clearance by homologous phagocytosis.
Acta histochemica (2014-12-17)
María Clara Carou, Paula Romina Cruzans, Alejandro Maruri, Juan Carlos Stockert, Daniel Marcelo Lombardo
ABSTRACT
Apoptosis is involved in many physiological processes of the ovary, such as recruitment of prenatal germ cells, follicular atresia, ovulation, and luteolysis. Based on the need for the involvement of phagocytic cells to achieve apoptosis clearance and that follicular atresia is triggered by weak apoptotic stimuli, we postulate that granulosa cells engullng apoptotic corpses (ACs) must carry out this macrophagic process. Since apoptosis was early defined in terms of morphological aspects, here we describe apoptosis induced by a GnRH analog (leuprolide acetate, LA) at histological level on bovine granulosa cells (primary culture, CPGB, and an established cell line, BGC-1). We observed two main types of apoptosis. In type A, the whole cell or most of it is compacted into a single large AC that is then engulfed by neighboring cells or simply detached. In type B, small portions of cells, either with or without nuclear material, become ACs that are also phagocytosed. Apoptosis and homologous phagocytosis were confirmed by TUNEL and immunocytochemistry for Bax and active caspase 3. Induction of apoptosis was significant in BGC-1 cells treated for 24 h with 100 nM LA. CPGB cells showed two types of response with different doses of LA. Fetal calf serum was necessary to find apoptosis induced by LA.
Product Description
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Luteinizing hormone releasing hormone, ≥97% (HPLC)
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
SAFC
L-Glutamine
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Luteinizing hormone releasing hormone salmon, ≥97% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%