Merck
  • Characterization of process-related impurities including forced degradation products of alogliptin benzoate and the development of the corresponding reversed-phase high-performance liquid chromatography method.
Characterization of process-related impurities including forced degradation products of alogliptin benzoate and the development of the corresponding reversed-phase high-performance liquid chromatography method.
Journal of separation science (2014-03-13)
YuXia Zhou, WenTao Zhou, LiLi Sun, QiaoGen Zou, Ping Wei, PingKai OuYang
ABSTRACT
The characterization of process-related impurities and forced degradants of alogliptin benzoate (Alb) in bulk drugs and a stability-indicating HPLC method for the separation and quantification of all the impurities were investigated. Alb was found to be unstable under acid and alkali stress conditions and two major degradation products (Imp-F and Imp-G) were observed. The optimum separation was achieved on Kromasil C18 (250 × 4.6 mm, 5 μm) using 0.1% perchloric acid (pH adjusted to 3.0 with triethylamine) and acetonitrile as a mobile phase in gradient mode. The proposed method was found to be stability indicating, precise, linear (0.10-75.0 μg/mL), accurate, sensitive, and robust for the quantitation of Alb and its process-related substances and degradation products. The structures of 11 impurities were characterized and confirmed by NMR spectroscopy, MS, and IR spectroscopy, and the most probable formation mechanisms of all impurities were proposed according to the synthesis route.
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Triethylamine, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis