Merck
  • Biochemical and hematological effects of acute and sub-acute administration to ethyl acetate fraction from the stem bark Scutia buxifolia Reissek in mice.
Biochemical and hematological effects of acute and sub-acute administration to ethyl acetate fraction from the stem bark Scutia buxifolia Reissek in mice.
Journal of ethnopharmacology (2014-04-08)
Andreia R H da Silva, Laís da R Moreira, Evelyne da S Brum, Mayara L de Freitas, Aline A Boligon, Margareth L Athayde, Silvane S Roman, Cinthia M Mazzanti, Ricardo Brandão
ABSTRACT
Scutia buxifolia is a native tree of Southern Brazil, Uruguay, and Argentina, which is popularly known as "coronilha" and it is used as a cardiotonic, antihypertensive and diuretic substance. The aim of this study was to assess the acute and sub-acute toxicity of the ethyl acetate fraction from the stem bark Scutia buxifolia in male and female mice. The toxicity studies were based on the guidelines of the Organization for Economic Cooperation and Development (OECD-guidelines 423 and 407). In an acute study, a single dose of 2000 mg/kg of Scutia buxifolia was administered orally to male and female mice. Mortality, behavioral changes, and biochemical and hematological parameters were evaluated. In the sub-acute study, Scutia buxifolia was administered orally to male and female mice at doses of 100, 200, and 400mg/kg/day for 28 days. Behavioral changes and biochemical, hematological, and histological analysis were evaluated. The acute administration of Scutia buxifolia did not cause changes in behavior or mortality. Male and female mice presented decreased levels of platelets. Female mice presented decreased levels of leukocytes. On the other hand, in a sub-acute toxicity study, we observed no behavioral changes in male or female mice. Our results demonstrated a reduction in glucose levels in male mice treated to 200 and 400mg/kg of Scutia buxifolia. Aspartate aminotransferase (ASAT) activity was increased by Scutia buxifolia at 400mg/kg in male mice. In relation to the hematological parameters, male mice presented a reduction in hemoglobin (HGB) and hematocrit (HCT) when treated to 400mg/kg of plant fraction. Female mice showed no change in these parameters. Histopathological examination of liver tissue showed slight abnormalities that were consistent with the biochemical variations observed. Scutia buxifolia, after acute administration, may be classified as safe (category 5), according to the OECD guide. However, the alterations observed, after sub-acute administration with high doses of ethyl acetate fraction from the stem bark Scutia buxifolia, suggest that repeated administration of this fraction plant can cause adverse hepatic, renal, and hematological effects.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Quercetin 3-glucoside, analytical standard
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Isoquercitrin, primary reference standard
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Chlorogenic acid, primary reference standard