Merck
  • Synthesis of lucifensin by native chemical ligation and characteristics of its isomer having different disulfide bridge pattern.
Synthesis of lucifensin by native chemical ligation and characteristics of its isomer having different disulfide bridge pattern.
Journal of peptide science : an official publication of the European Peptide Society (2014-06-13)
Stancho Stanchev, Zbigniew Zawada, Lenka Monincová, Lucie Bednárová, Jiřina Slaninová, Vladimír Fučík, Václav Čeřovský
ABSTRACT
The antimicrobial 40-amino-acid-peptide lucifensin was synthesized by native chemical ligation (NCL) using N-acylbenzimidazolinone (Nbz) as a linker group. NCL is a method in which a peptide bond between two discreet peptide chains is created. This method has been applied to the synthesis of long peptides and proteins when solid-phase synthesis is imcompatible. Two models of ligation were developed: [15+25] Ala-Cys and [19+21] His-Cys. The [19+21] His-Cys method gives lower yield because of the lower stability of 18-peptide-His-Nbz-CONH2 peptide, as suggested by density functional theory calculation. Acetamidomethyl-deprotection and subsequent oxidation of the ligated linear lucifensin gave a mixture of lucifensin isomers, which differed in the location of their disulfide bridges only. The dominant isomer showed unnatural pairing of cysteines [C1-6], [C3-5], and [C2-4], which limits its ability to form α-helical structure. The activity of isomeric lucifensin toward Bacillus subtilis, Staphylococcus aureus, and Micrococcus luteus was lower than that of the natural lucifensin. The desired product native lucifensin was prepared from this isomer using a one-pot reduction with dithiotreitol and subsequent air oxidation in slightly alkaline medium.
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
N-Ethyldiisopropylamine, ≥98.0%
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Fmoc N-hydroxysuccinimide ester, ≥98.0% (HPLC)
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Supelco
Piperidine, analytical standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Piperidine solution, suitable for peptide synthesis, 20% in DMF
Supelco
Acetonitrile, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer