Merck
  • Mixed ligand copper(II) complexes of 1,10-phenanthroline with tridentate phenolate/pyridyl/(benz)imidazolyl Schiff base ligands: covalent vs non-covalent DNA binding, DNA cleavage and cytotoxicity.
Mixed ligand copper(II) complexes of 1,10-phenanthroline with tridentate phenolate/pyridyl/(benz)imidazolyl Schiff base ligands: covalent vs non-covalent DNA binding, DNA cleavage and cytotoxicity.
Journal of inorganic biochemistry (2014-09-10)
Chandrasekaran Rajarajeswari, Mani Ganeshpandian, Mallayan Palaniandavar, Anvarbatcha Riyasdeen, Mohammad Abdulkadher Akbarsha
ABSTRACT
A series of copper(II) complexes of the types [Cu(L)(phen)](ClO4) 1-2, where HL is a tridentate ligand with two nitrogen and one oxygen donor atoms (2NO) such as 2-(2-(1H-benzimidazol-2-yl)ethyliminomethyl)phenol (HL1) and 2-(2-(1H-benzimidazol-2-yl)ethyl-imino)methyl)-4-methylphenol (HL2), phen is 1,10-phenanthroline and [Cu(L)(phen)](ClO4)23-6, where L is a tridentate ligand with three nitrogen donor atoms (3N) such as (2-pyridin-2-ylethyl)pyridin-2-ylmethyleneamine (L3), 2-(1H-benzimidazol-2-yl)ethyl)-pyridin-2-yl-methyleneamine (L4), 2-(1H-benzimidazol-2-yl)ethyl)(1H-imidazol-2-ylmethylene)-amine (L5) and 2-(1H-benzimidazol-2-yl)ethyl)(4,4a-dihydroquinolin-2-ylmethylene)amine (L6), has been isolated and characterized by different spectral techniques. In single crystal X-ray structures, 1 possesses square pyramidal distorted trigonal bipyramidal (SPDTBP), geometry whereas 3 and 4 possess trigonal bipyramidal distorted square pyramidal (TBDSP) geometry. UV-Vis and fluorescence spectral studies reveal that the complexes 1-6 bind non-covalently to calf thymus DNA more strongly than the corresponding covalently bound chlorido complexes [Cu(2NO)Cl] 1a-2a and [Cu(3N)Cl2] 3a-6a. On prolonged incubation, all the complexes 1-6 exhibit double strand cleavage of supercoiled (SC) plasmid DNA in the absence of an activator. Also, they exhibit cytotoxicity against human breast cancer cell lines (HBL-100) more potent than their corresponding chlorido complexes 1a-6a, and have the potential to act as efficient cytotoxic drugs.
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Copper(II) perchlorate hexahydrate, 98%
Sigma-Aldrich
2-Quinolinecarboxaldehyde, 97%
Sigma-Aldrich
2-Imidazolecarboxaldehyde, 97%
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
2-Pyridinecarboxaldehyde, 99%
Sigma-Aldrich
2-Hydroxy-5-methylbenzaldehyde, 98%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%