Merck
  • Alleviating waiting impulsivity and perseverative responding by μ-opioid receptor antagonism in two inbred mouse strains.
Alleviating waiting impulsivity and perseverative responding by μ-opioid receptor antagonism in two inbred mouse strains.
Psychopharmacology (2014-11-09)
Sandra Sanchez-Roige, Tamzin L Ripley, David N Stephens
ABSTRACT
Recent evidence has implicated the opioid system in exaggerated ethanol consumption and impulsivity deficits. The opioid receptor antagonist naltrexone (NTX) has proven efficient in reducing alcohol consumption; however, its role on impulsive behaviour is not fully characterised. The aim of this study was to investigate the effects of NTX on two measures of impulsive behaviour in two inbred mouse strains that differ in ethanol preference and impulsive phenotype. Two separate groups of C57BL/6J (B6, n = 24) and DBA2/J (D2, n = 24) male mice were exposed to intermittent ethanol (IEE; 2 g/kg) during early (PND 30-45, IEE_Early) or late (PND 45-60, IEE_Late) adolescence or the respective saline control. The ability of NTX (10 mg/kg) alone, or co-administered with ethanol (0.5 g/kg), to diminish waiting impulsivity in the five-choice serial reaction time task (5-CSRTT), or improve decision-making in a mouse version of the Iowa Gambling Task (mIGT), was examined in adulthood. In the 5-CSRTT, NTX diminished impulsivity in both strains of mice, irrespective of previous ethanol experience. In the mIGT, NTX failed to alter risky decision-making but decreased perseverative responding. Blocking the actions of endogenous opioids may attenuate waiting impulsivity, in addition to alleviating perseverative responding. In a broader context, μ-opiate antagonism may be of potential interest for impulse-control disorders.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naltrexone hydrochloride
Naltrexone hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology