Merck
  • Synthesis of lysozyme-metallacarborane conjugates and the effect of boron cluster modification on protein structure and function.
Synthesis of lysozyme-metallacarborane conjugates and the effect of boron cluster modification on protein structure and function.
Chembiochem : a European journal of chemical biology (2015-01-16)
Konrad Kowalski, Tomasz Goszczyński, Zbigniew J Leśnikowski, Janusz Boratyński
ABSTRACT
Two complementary methods, "in solution" and "in solid state", for the synthesis of lysozyme modified with metallacarborane (cobalt bis(dicarbollide), Co(C2 B9 H11 )2 (2-) ) were developed. As metallacarborane donors, oxonium adducts of cobalt bis(dicarbollide) and 1,4-dioxane or tetrahydropyran were used. The physicochemical and biochemical properties of the obtained lysozyme-metallacarborane conjugates were studied for changes in secondary and tertiary structure, aggregation behavior, and biological activity. Only minor changes in primary, secondary, and tertiary protein structure were observed, caused by the single substitution of metallacarborane on lysozyme. However, the modification produced significant changes in lysozyme enzymatic activity and a tendency toward time- and temperature-dependent aggregation.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
4-Aminosalicylic acid, 99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard