Merck
  • Hematopoietic and mesenchymal stem cells: polymeric nanoparticle uptake and lineage differentiation.
Hematopoietic and mesenchymal stem cells: polymeric nanoparticle uptake and lineage differentiation.
Beilstein journal of nanotechnology (2015-03-31)
Ivonne Brüstle, Thomas Simmet, Gerd Ulrich Nienhaus, Katharina Landfester, Volker Mailänder
ABSTRACT
The combination of stem cell therapy and nanoparticles promises to enhance the effect of cellular therapies by using nanocarriers as drug delivery devices to guide the further differentiation or homing of stem cells. The impact of nanoparticles on primary cell types remains much more elusive as most groups study the nanoparticle-cell interaction in malignant cell lines. Here, we report on the influence of polymeric nanoparticles on human hematopoietic stem cells (hHSCs) and mesenchymal stem cells (hMSCs). In this study we systematically investigated the influence of polymeric nanoparticles on the cell functionality and differentiation capacity of hHSCs and hMSCs to obtain a deeper knowledge of the interaction of stem cells and nanoparticles. As model systems of nanoparticles, two sets of either bioinert (polystyrene without carboxylic groups on the surface) or biodegradable (PLLA without magnetite) particles were analyzed. Flow cytometry and microscopy analysis showed high uptake rates and no toxicity for all four tested particles in hMSCs and hHSCs. During the differentiation process, the payload of particles per cell decreased. The PLLA-Fe particle showed a significant increase in the IL-8 release in hMSCs but not in hHSCs. We assume that this is due to an increase of free intracellular iron ions but obviously also depends on the cell type. For hHSCs and hMSCs, lineage differentiation into erythrocytes, granulocytes, and megakaryocytes or adipocytes, osteocytes and chondrocytes, was not influenced by the particles when analyzed with lineage specific cluster of differentiation markers. On the other hand qPCR analysis showed significant changes in the expression of some (but not all) investigated lineage markers for both primary cell types.
Product Description
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene blue, certified by the BSC