Merck
  • Structure-based design of phthalimide derivatives as potential cyclooxygenase-2 (COX-2) inhibitors: anti-inflammatory and analgesic activities.
Structure-based design of phthalimide derivatives as potential cyclooxygenase-2 (COX-2) inhibitors: anti-inflammatory and analgesic activities.
European journal of medicinal chemistry (2015-01-01)
Amer M Alanazi, Adel S El-Azab, Ibrahim A Al-Suwaidan, Kamal Eldin H ElTahir, Yousif A Asiri, Naglaa I Abdel-Aziz, Alaa A-M Abdel-Aziz
ABSTRACT
A group of 30 cyclic imides (1-10a-c) was designed for evaluation as a selective COX-2 inhibitor and investigated in vivo for anti-inflammatory and analgesic activities. Compounds 6a, 6b, 7a and 7b exhibit optimal COX-2 inhibitory potency (IC50 = 0.18, 0.24, 0.28 and 0.36 μM; respectively) and selectivity index (SI) range of 363-668. In vitro COX-1/COX-2 inhibition structure-activity studies identified compound 6a as a highly potent (IC50 = 0.18 μM), and an extremely selective [COX-2 (SI) = 668] comparable to celecoxib [COX-2 (SI) > 384], COX-2 inhibitor that showed superior anti-inflammatory activity (ED50 = 54.0 mg/kg) relative to diclofenac (ED50 = 114 mg/kg). Molecular Docking study of the synthesized compound 6a into the active site of COX-2 revealed a similar binding mode to SC-558, a selective COX-2 inhibitor. Docking study showed that the methoxy moeities of 6a inserted deep inside the 2°-pocket of the COX-2 active site, where the O-atoms of such groups underwent an H-bonding interaction with His(90) (3.02 Å), Arg(513) (1.94, 2.83 Å), and Gln(192) (3.25 Å).
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
1,3-Propylene sulfite, 99%