Merck
  • Pressurized liquid extraction and liquid chromatography-tandem mass spectrometry applied to determine iodinated X-ray contrast media in sewage sludge.
Pressurized liquid extraction and liquid chromatography-tandem mass spectrometry applied to determine iodinated X-ray contrast media in sewage sludge.
Analytica chimica acta (2014-08-31)
S Echeverría, F Borrull, E Pocurull, N Fontanals
ABSTRACT
Pressurized liquid extraction (PLE) has been successfully applied for the first time to the extraction of five iodinated X-ray contrast media from sludge. Once optimized all PLE parameters, the extract has been analyzed by liquid chromatography-tandem mass spectrometry, being the method developed sensible enough to reach limit of quantifications (LOQs) of 25 μg kg(-1) (d.w.). The developed method has been applied to the analysis of sludge from urban sewage treatment plants and although some compounds such as iopromide, diatrizoic acid and iopamidol have been identified, their concentrations have been lower than their LOQs.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Methanol, analytical standard
Supelco
Acetone, analytical standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard