Merck
  • Photocatalytic degradation of methylene blue with a nanocomposite system: synthesis, photocatalysis and degradation pathways.
Photocatalytic degradation of methylene blue with a nanocomposite system: synthesis, photocatalysis and degradation pathways.
Physical chemistry chemical physics : PCCP (2015-01-23)
Shengjie Xia, Lianyang Zhang, Guoxiang Pan, Pingping Qian, Zheming Ni
ABSTRACT
Three different composites, including a calcined FeOOH supported ZnAl layered double hydroxide (FeOOH-LDO), a calcined ZnAl layered double hydroxide (ZnAl-LDO) and a calcined ZnFeAl layered double hydroxide (ZnFeAl-LDO), were synthesized via a sol-gel method, and their activity for the visible light photocatalytic degradation of methylene blue (MB) was studied. The composites were characterized by PXRD, SEM, and BET techniques, confirming the formation of highly crystalline structures. The activity performance of MB degradation was in the following order: FeOOH-LDO (∼95%) > ZnFeAl-LDO (∼60%) > ZnAl-LDO (∼23%). In addition, a possible photocatalytic degradation reaction mechanism for MB was also proposed. Moreover, the frontier electron densities on the atoms of MB were calculated, which were in satisfactory agreement with the postulated mechanism.
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene blue, certified by the BSC