Merck
Pueraria mirifica leaves, an alternative potential isoflavonoid source.
Bioscience, biotechnology, and biochemistry (2014-07-19)
Jutarmas Jungsukcharoen, Binar Asrining Dhiani, Wichai Cherdshewasart, Nawaporn Vinayavekhin, Polkit Sangvanich, Chuenchit Boonchird
ABSTRACT
We investigated the major leaf isoflavonoid contents of Pueraria mirifica from three different cultivars (PM-III, PM-IV, and PM-V) using reverse RP-HPLC analysis. The proportions and net levels of puerarin, daidzin, genistin, and daidzein in P. mirifica leaves were found to depend on the plant cultivar and to correlate with cultivation temperature and rainfall amount. The crude leaf-extracts were tested using the Yeast Estrogen Screen (YES) assay with both human estrogen receptors (hERα and hERβ). Their estrogenic activity was higher when determined by the YES system containing hERβ than that with hERα and was also higher when the Δsnq2 than the wildtype yeast was employed. The results open the possibility of selecting and cultivating certain P. mirifica cultivars at a farm scale to produce a sufficient supply of leaf material to act as a starting source for the commercial scale extraction of these major isoflavonoids.
Product Description
Supelco
Genistein, analytical standard
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Supelco
Daidzein, analytical standard
Sigma-Aldrich
Daidzein, ≥98%, synthetic