Merck
  • Evaluation of neuronal protective effects of xanthine oxidoreductase inhibitors on severe whole-brain ischemia in mouse model and analysis of xanthine oxidoreductase activity in the mouse brain.
Evaluation of neuronal protective effects of xanthine oxidoreductase inhibitors on severe whole-brain ischemia in mouse model and analysis of xanthine oxidoreductase activity in the mouse brain.
Neurologia medico-chirurgica (2015-03-07)
Go Suzuki, Ken Okamoto, Teruo Kusano, Yoko Matsuda, Akira Fuse, Hiroyuki Yokota
ABSTRACT
Global cerebral ischemia and reperfusion (I/R) often result in high mortality. Free radicals play an important role in global cerebral I/R. Xanthine oxidoreductase (XOR) inhibitors, such as allopurinol, have been reported to protect tissues from damage caused by reactive oxygen species (ROS) by inhibiting its production through XOR inhibition. The recently introduced XOR inhibitor febuxostat, which is a more potent inhibitor than allopurinol, is expected to decrease free radical production more effectively. Here, we analyzed the effects of allopurinol and febuxostat in decreasing global severe cerebral I/R damage in mice. Mice were divided into three groups: a placebo group, an allopurinol group, and a febuxostat group. Pathological examinations, which were performed in each group in the CA1 and CA2 regions of the hippocampus 4 days after I/R surgery, revealed that there was a decrease in the number of neuronal cells in the 14-min occlusion model in both regions and that drugs that were administered to prevent this damage were not effective. The enzymatic activity was extremely low in the mouse brain, and XOR could not be detected in the nonischemic and ischemic mice brains with western blot analyses. Thus, one of the reasons for the decreased effectiveness of XOR inhibitors in controlling severe whole-brain ischemia in a mouse model was the low levels of expression of XOR in the mouse brain.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Oxypurinol, ≥98% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard
USP
Allopurinol, United States Pharmacopeia (USP) Reference Standard
Allopurinol, European Pharmacopoeia (EP) Reference Standard