  • Search Results
  • Development of an immunoaffinity solid phase microextraction method for the identification of penicillin binding protein 2a.
Development of an immunoaffinity solid phase microextraction method for the identification of penicillin binding protein 2a.
Journal of chromatography. A (2014-09-04)
Yuhong Liu, Heather Lord, Małgorzata Maciążek-Jurczyk, Sukhsagar Jolly, Mohamed Asim Hussain, Janusz Pawliszyn
ABSTRACT
Methicillin resistant Staphylococcus aureus (MRSA) is a bacterium that is resistant to many antibiotics. Resistance to methicillin is related to production of penicillin binding protein 2a (PBP2a). The currently presented research involves the development of antibody-linked immunoaffinity solid phase microextraction (SPME) sorbents characterized from several aspects that can identify protein PBP2a. The on-sorbent binding constant Kd of monoclonal anti-PBP2a antibody for its antigen protein PBP2a was determined to be 4×10(-10) M. This value was obtained on the basis of the binding curve determined by selective extraction of its antigen PBP2a at different concentrations. The concentration of PBP2a captured by immunoaffinity sorbents was as low as 10ng/mL; lower concentrations could not be tested due to the sensitivity limitation of the LC-MS/MS system available. Surface density was estimated at 59 ng antibody/cm(2). To reduce non-specific binding, especially when the antigen is a protein, bovine serum albumin (BSA) was used to pretreat surfaces. The established immunoaffinity platform technology is expected to provide insights into the development of a practical, specific, sensitive and accurate assay for in vitro and in vivo diagnostics of MRSA.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Ethanol, for residue analysis
Supelco
Acetonitrile, analytical standard
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O