Merck
  • Dasatinib (BMS-35482) interacts synergistically with docetaxel, gemcitabine, topotecan, and doxorubicin in ovarian cancer cells with high SRC pathway activation and protein expression.
Dasatinib (BMS-35482) interacts synergistically with docetaxel, gemcitabine, topotecan, and doxorubicin in ovarian cancer cells with high SRC pathway activation and protein expression.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society (2014-01-11)
Angeles Alvarez Secord, Deanna Teoh, Jingquan Jia, Andrew B Nixon, Lisa Grace, David J Adams, Susan K Murphy
ABSTRACT
This study aimed to explore the activity of dasatinib in combination with docetaxel, gemcitabine, topotecan, and doxorubicin in ovarian cancer cells. Cells with previously determined SRC pathway and protein expression (SRC pathway/SRC protein IGROV1, both high; SKOV3, both low) were treated with dasatinib in combination with the cytotoxic agents. SRC and paxillin protein expression were determined pretreatment and posttreatment. Dose-response curves were constructed, and the combination index (CI) for drug interaction was calculated. In the IGROV1 cells, dasatinib alone reduced phospho-SRC/total SRC 71% and p-paxillin/t-paxillin ratios 77%. Phospho-SRC (3%-33%; P = 0.002 to 0.04) and p-paxicillin (6%-19%; P = 0.01 to 0.05) levels were significantly reduced with dasatinib in combination with each cytotoxic agent. The combination of dasatinib and docetaxel, gemcitabine, or topotecan had a synergistic antiproliferative effect (CI, 0.49-0.68), whereas dasatinib combined with doxorubicin had an additive effect (CI, 1.08).In SKOV3 cells, dasatinib resulted in less pronounced reductions of phospho-SRC/total SRC (49%) and p-paxillin/t-paxillin (62%). Phospho-SRC (18%; P < 0.001) and p-paxillin levels (18%; P = 0.001; 9%; P = 0.007) were significantly decreased when dasatinib was combined with docetaxel and topotecan (p-paxillin only). Furthermore, dasatinib combined with the cytotoxics in the SKOV3 cells produced an antagonistic interaction on the proliferation of these cells (CI, 1.49-2.27). Dasatinib in combination with relapse chemotherapeutic agents seems to interact in a synergistic or additive manner in cells with high SRC pathway activation and protein expression. Further evaluation of dasatinib in combination with chemotherapy in ovarian cancer animal models and exploration of the use of biomarkers to direct therapy are warranted.
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Anhydrous Docetaxel, European Pharmacopoeia (EP) Reference Standard
USP
Docetaxel, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Topotecan hydrochloride hydrate, ≥98% (HPLC and enzymatic)