Merck
  • Microbiological and physicochemical analysis of yateí (Tetragonisca angustula) honey for assessing quality standards and commercialization.
Microbiological and physicochemical analysis of yateí (Tetragonisca angustula) honey for assessing quality standards and commercialization.
Revista Argentina de microbiologia (2015-01-13)
Amada B Pucciarelli, María E Schapovaloff, Silvana Kummritz, Isabel A Señuk, Luis A Brumovsky, Andrea M Dallagnol
ABSTRACT
Due to the interest in the production and trading of yateí (Tetragonisca angustula) honey in the province of Misiones, Argentina, in this work we assessed microbiological and physicochemical parameters in order to contribute to the elaboration of standards for quality control and promote commercialization. Results showed that yateí honey samples had significantly different microbiological and physicochemical characteristics in comparison to established quality standards for Apis mellifera honey. Thus, we observed that values for pH (3.72), glucose (19.01 g/100g) and fructose (23.74 g/100g) were lower than A. mellifera quality standards, while acidity (79.42 meq/kg), moisture (24%), and mould and yeast count (MY) (3.02 log CFU/g) were higher. The acid content was correlated with glucose (R2=0.75) and fructose (R2=0.68) content, and also with mould and yeast counts (R2=0.45) to a lesser extent. The incidence of microorganisms in yateí honey samples reached 42.85% and 39% for Clostridium sulfite-reducers and Bacillus spp., respectively. No C. botulinum or B. cereus cells were detected. Enterococcus spp. and Staphylococcus spp. incidence was similar (ca. 7.14%), whereas Escherichia coli and Salmonella spp. were not detected. We conclude that the microbiological and physicochemical properties of yateí honey are different from those of A. mellifera honey; hence, different quality standards could be implemented to promote its commercialization.
Product Description
Sucrose, European Pharmacopoeia (EP) Reference Standard
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
USP
Fructose, United States Pharmacopeia (USP) Reference Standard