Merck
  • Latent heparanase facilitates VLA-4-mediated melanoma cell binding and emerges as a relevant target of heparin in the interference with metastatic progression.
Latent heparanase facilitates VLA-4-mediated melanoma cell binding and emerges as a relevant target of heparin in the interference with metastatic progression.
Seminars in thrombosis and hemostasis (2015-02-16)
Ursula Gerber, Sebastian G Hoß, Anna Shteingauz, Eva Jüngel, Bastian Jakubzig, Neta Ilan, Roman Blaheta, Martin Schlesinger, Israel Vlodavsky, Gerd Bendas
ABSTRACT
Heparanase is an endo-β-glucuronidase that enzymatically cleaves heparan sulfates (HS) and heparan sulfate proteoglycan (HSPG) structures. Heparanase expression levels by tumors were correlated with cell invasion, angiogenic activity, and poor prognosis. Heparanase can also possess pro-tumorigenic effects independent of its enzymatic activity. Using human melanoma MV3 cells, we demonstrate that latent heparanase activates in a tightly temporary-regulated manner the binding function of the integrin very late antigen-4 (VLA-4), an important component in the metastatic spread of melanoma cells. shRNA-mediated knockdown of syndecan-4 (SDC-4) indicated that this proteoglycan is the key element to convey heparanase binding via focal adhesion complex formation, detected by vinculin staining, to an upregulated VLA-4 binding function. This inside-out signaling pathway of VLA-4 involved activated FAK and Akt, but apparently not PKCα/δ. VLA-4, however, appears representative of other integrins which together impact the heparanase/integrin activation axis in tumorigenicity. Biosensor measurements provided an insight as to how heparin can interfere with this activation process. While low-molecular-weight heparin (LMWH) cannot replace heparanase bound to SDC-4, LMWH can compete with SDC-4 binding of heparanase. Since blockade of heparanase by LMWH has functional consequences for reduced VLA-4 binding, latent heparanase appears as a novel, so far unnoticed target of heparin, underlying its antimetastatic activity.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Cerium(IV) oxide-samarium doped, <0.5 μm particle size, powder, contains 20 mol % samarium as dopant
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Cyanuric chloride, 99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder