Merck
  • Ion mobility spectrometry evaluation of cocaine occupational exposure in forensic laboratories.
Ion mobility spectrometry evaluation of cocaine occupational exposure in forensic laboratories.
Talanta (2014-08-28)
Sergio Armenta, Miguel de la Guardia, Manel Alcalà, Marcelo Blanco, Clara Perez-Alfonso, Nieves Galipienso
ABSTRACT
An approach, based on ion mobility spectrometry (IMS) has been developed for the control of cocaine in air of the breathing zone of operators, in laboratory surfaces and in nasal mucus of employees to evaluate cocaine exposure in a forensic laboratory. The analytical methodology has been validated in terms of accuracy, precision and limits of detection and results obtained were statistically comparable with those obtained by liquid chromatography. Cocaine concentration in laboratory air increases from 100 ± 35 ng m(-3) of a normal day to 10,000 ng m(-3) during the manipulation of cocaine seizures. The occupational exposure limit (OEL) for cocaine has not been established which difficult the evaluation of the health effects of continuous exposition to very small doses of cocaine. Cocaine was also found in almost all the analyzed sample surfaces and also was found in nasal mucus of the police officers that were present during the manipulation of cocaine seizures without using a face mask. In summary, cocaine concentrations could present a health hazard to the employees and therefore warrants remediation and some modifications of the manipulation operations have been proposed.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ultrapure Acetonitrile
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%