  • Search Results
  • Cell response to single-walled carbon nanotubes in hybrid porous collagen sponges.
Cell response to single-walled carbon nanotubes in hybrid porous collagen sponges.
Colloids and surfaces. B, Biointerfaces (2014-12-30)
Hongli Mao, Naoki Kawazoe, Guoping Chen
ABSTRACT
Three-dimensional (3D) porous collagen sponges incorporated with single-walled carbon nanotubes (SWCNTs) were prepared and used for 3D culture of bovine articular chondrocytes (BACs). The pore structures of the sponges were controlled by using ice particulates as a porogen material. The responses of cells to SWCNTs were investigated in this 3D cell culture system by evaluation of cell functions and cellular uptake of SWCNTs. The results showed that cells adhered and spatially distributed in the porous sponges. The incorporation of SWCNTs in the porous sponges promoted cell proliferation and production of sulfated glycosaminoglycans (sGAG). Confocal Raman imaging revealed that SWCNTs could be internalized by cells. The hybrid porous sponges not only provided nanostructured pore surfaces to facilitate cell proliferation and extracellular matrix (ECM) secretion but also supplied nanomaterials for cellular uptake which may be useful for biomedical applications.
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
SAFC
Acetic acid, glacial
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
SAFC
L-Cysteine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
L-Cysteine, certified reference material, TraceCERT®
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard