Merck
  • Generation of induced pluripotent stem cells without genetic defects by small molecules.
Generation of induced pluripotent stem cells without genetic defects by small molecules.
Biomaterials (2014-12-06)
Hang-Soo Park, Insik Hwang, Kyung-Ah Choi, Hyesun Jeong, Ji-Yun Lee, Sunghoi Hong
ABSTRACT
The generation of induced pluripotent stem cells (iPSCs) often causes genetic and epigenetic defects, which may limit their clinical applications. Here, we show that reprogramming in the presence of small molecules preserved the genomic stability of iPSCs by inhibiting DNA double-strand breaks (DSBs) and activating Zscan4 gene. Surprisingly, the small molecules protected normal karyotype by facilitating repair of the DSBs that occurred during the early reprogramming process and long-term culture of iPSCs. The stemness and cell growth of iPSCs(+) were normally sustained with high expression of pluripotency genes compared that of iPSCs(-). Moreover, small molecules maintained the differentiation potential of iPSCs(+) for the three germ layers, whereas it was lost in iPSCs(-). Our results demonstrate that the defined small molecules are potent factors for generation of high quality iPSCs with preservation of genomic integrity by facilitating the reprogramming process.
Product Description
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
2-Propylpentanoic acid
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Valproic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)