Merck
  • UHRF1 depletion suppresses growth of gallbladder cancer cells through induction of apoptosis and cell cycle arrest.
UHRF1 depletion suppresses growth of gallbladder cancer cells through induction of apoptosis and cell cycle arrest.
Oncology reports (2014-04-24)
Yiyu Qin, Jiandong Wang, Wei Gong, Mingdi Zhang, Zhaohui Tang, Jun Zhang, Zhiwei Quan
ABSTRACT
Ubiquitin-like containing PHD and RING finger domains 1 (UHRF1), overexpressed in various human malignancies, functions as an important regulator in cell proliferation and epigenetic regulation. Depletion of UHRF1 has shown potential antitumor activities in several types of cancer. However, the role of UHRF1 in gallbladder cancer (GBC) has not been investigated. RT-PCR, western blotting and immunohistochemistry were performed to examine UHRF1 expression at mRNA and protein levels in GBC tissues and cell lines. UHRF1 siRNA and UHRF1 shRNA were used to deplete the expression of UHRF1. The results showed that UHRF1 was overexpressed in GBC and its expression correlated with advanced TNM stage and presence of lymph node metastasis. UHRF1 depletion in GBC-SD and NOZ cells markedly inhibited proliferation, migration in vitro and the ability of these cells to form tumors in vivo. UHRF1 depletion upregulated the expression of PML and triggered extrinsic and intrinsic apoptotic pathways by promoting the expression of FasL/FADD, bax, cytosolic cytochrome c, cleaved caspase-8, -9 and -3 and cleaved PRAP and by suppressing bcl-2 expression in GBC-SD and NOZ cells. In addition, UHRF1 depletion induced cell cycle arrest at G1/S transition by inducing p21 in a p53-independent manner in GBC-SD and NOZ cells. Our findings suggest that UHRF1 is involved in the proliferation and migration of GBC cells and may serve as a biomarker or even a therapeutic target for GBC.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Uhrf1
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human UHRF1
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)