  • Search Results
  • NMR-based molecular ruler for determining the depth of intercalants within the lipid bilayer. Part IV: studies on ketophospholipids.
NMR-based molecular ruler for determining the depth of intercalants within the lipid bilayer. Part IV: studies on ketophospholipids.
Chemistry and physics of lipids (2014-07-30)
Michal Afri, Carmit Alexenberg, Pinchas Aped, Efrat Bodner, Sarit Cohen, Michal Ejgenberg, Shlomi Eliyahu, Pessia Gilinsky-Sharon, Yifat Harel, Miriam E Naqqash, Hani Porat, Ayala Ranz, Aryeh A Frimer
ABSTRACT
In our companion paper, we described the preparation and intercalation of two homologous series of dicarbonyl compounds, methyl n-oxooctadecanoates and the corresponding n-oxooctadecanoic acids (n=4-16), into DMPC liposomes. (13)C NMR chemical shift of the various carbonyls was analyzed using an E(T)(30) solvent polarity-chemical shift correlation table and the corresponding calculated penetration depth (in Å). An iterative best fit analysis of the data points revealed an exponential correlation between E(T)(30) micropolarity and the penetration depth (in Å) into the liposomal bilayer. However, this study is still incomplete, since the plot lacks data points in the important area of moderately polarity, i.e., in the E(T)(30) range of 51-45.5 kcal/mol. To correct this lacuna, a family of ketophospholipids was prepared in which the above n-oxooctadecanoic acids were attached to the sn-2 position of a phosphatidylcholine with a palmitic acid chain at sn-1. To assist in assignment and detection several derivatives were prepared (13)C-enriched in both carbonyls. The various homologs were intercalated into DMPC liposomes and give points specifically in the missing area of the previous polarity-penetration correlation graph. Interestingly, the calculated exponential relationship of the complete graph was essentially the same as that calculated in the companion paper based on the methyl n-oxooctadecanoates and the corresponding n-oxooctadecanoic acids alone. The polarity at the midplane of such DMPC systems is ca. 33 kcal/mol and is not expected to change very much if we extend the lipid chains. This paper concludes with a chemical ruler that maps the changing polarity experienced by an intercalant as it penetrates the liposomal bilayer.
Product Description
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
Carbon nanofibers, graphitized (iron-free), composed of conical platelets, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon - Vitreous, foil, 10mm disks, thickness 0.2mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foil, 8mm disks, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%