  • Search Results
  • Plasma metabonomic profiling of lumbar disc herniation and its traditional Chinese medicine subtypes in patients by using gas chromatography coupled with mass spectrometry.
Plasma metabonomic profiling of lumbar disc herniation and its traditional Chinese medicine subtypes in patients by using gas chromatography coupled with mass spectrometry.
Molecular bioSystems (2014-08-22)
Letian Shan, Fei Liao, Hongting Jin, Fusheng Ye, Peijian Tong, Luwei Xiao, Jia Zhou, Chengliang Wu
ABSTRACT
Lumbar disc herniation (LDH) is a commonly occurring disease, threatening human health and life quality. Lack of a gold standard of diagnosis has hindered the efficiency and efficacy of clinical therapy against LDH. Traditional Chinese medicine (TCM) has provided an experience-based but subjective diagnosis system for LDH, demanding objective evidence and explanation. In this study, we adopted a metabonomics approach using gas chromatography-mass spectrometry (GC-MS) to profile metabolic characteristics of LDH and its TCM subtypes. Plasma samples of 41 LDH patients and 25 healthy controls were collected. LDH patients were classified into two main subtypes, the reality syndrome and deficiency syndrome, according to TCM theory. By using multivariate statistical analysis and metabolism network analysis, we found diverse perturbations of metabolites in amino acid metabolism and carbohydrate metabolism, in which the amino acids (glutamic acid, aspartic acid, glycine, etc.) were up-regulated and a key carbohydrate metabolite (glucose 1-phosphate) was down-regulated. Few differences were found between the two TCM subtypes. Our findings reveal the metabolic disorders of LDH for the first time and demonstrate the feasibility of the metabonomics approach for LDH research but not for its TCM subtypes.
Product Description
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Methoxylamine hydrochloride solution, 25-30 wt. % in H2O
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material