Merck
  • Integrin-linked kinase plays a key role in the regulation of angiotensin II-induced renal inflammation.
Integrin-linked kinase plays a key role in the regulation of angiotensin II-induced renal inflammation.
Clinical science (London, England : 1979) (2014-01-05)
Matilde Alique, Esther Civantos, Elsa Sanchez-Lopez, Carolina Lavoz, Sandra Rayego-Mateos, Raquel Rodrigues-Díez, Ana Belén García-Redondo, Jesús Egido, Alberto Ortiz, Diego Rodríguez-Puyol, Manuel Rodríguez-Puyol, Marta Ruiz-Ortega
ABSTRACT
ILK (integrin-linked kinase) is an intracellular serine/threonine kinase involved in cell-matrix interactions. ILK dysregulation has been described in chronic renal disease and modulates podocyte function and fibrosis, whereas data about its role in inflammation are scarce. AngII (angiotensin II) is a pro-inflammatory cytokine that promotes renal inflammation. AngII blockers are renoprotective and down-regulate ILK in experimental kidney disease, but the involvement of ILK in the actions of AngII in the kidney has not been addressed. Therefore we have investigated whether ILK signalling modulates the kidney response to systemic AngII infusion in wild-type and ILK-conditional knockout mice. In wild-type mice, AngII induced an inflammatory response, characterized by infiltration of monocytes/macrophages and lymphocytes, and up-regulation of pro-inflammatory factors (chemokines, adhesion molecules and cytokines). AngII activated several intracellular signalling mechanisms, such as the NF-κB (nuclear factor κB) transcription factor, Akt and production of ROS (reactive oxygen species). All these responses were prevented in AngII-infused ILK-deficient mice. In vitro studies characterized further the mechanisms regulating the inflammatory response modulated by ILK. In cultured tubular epithelial cells ILK blockade, by siRNA, inhibited AngII-induced NF-κB subunit p65 phosphorylation and its nuclear translocation. Moreover, ILK gene silencing prevented NF-κB-related pro-inflammatory gene up-regulation. The results of the present study demonstrate that ILK plays a key role in the regulation of renal inflammation by modulating the canonical NF-κB pathway, and suggest a potential therapeutic target for inflammatory renal diseases.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Dihydroethidium, ≥95%
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting human ILK
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α from rat, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture