Merck
  • Monofunctional platinum(II) complexes with potent tumor cell growth inhibitory activity: the effect of a hydrogen-bond donor/acceptor N-heterocyclic ligand.
Monofunctional platinum(II) complexes with potent tumor cell growth inhibitory activity: the effect of a hydrogen-bond donor/acceptor N-heterocyclic ligand.
ChemMedChem (2014-05-07)
Nicola Margiotta, Salvatore Savino, Valentina Gandin, Christine Marzano, Giovanni Natile
ABSTRACT
In this paper we investigate the possibility of further increase the role of the N-donor aromatic base in antitumor Hollis-type compounds by conferring the possibility to act as a hydrogen-bond donor/acceptor. Therefore, we synthesized the Pt(II) complex cis-[PtCl(NH3 )2 (naph)]NO3 (1) containing the 1,8-naphthyridine (naph) ligand. The naphthyridine ligand is generally monodentate, and the second nitrogen atom can act as H-bond donor/acceptor depending upon its protonation state. The possibility of forming such an H-bond could be crucial in the interaction of the drug with DNA or proteins. Apart from the synthesis of the compound, in this study we evaluated its in vitro antitumor activity in a wide panel of tumor cell lines, also including cells selected for their sensitivity/resistance to oxaliplatin, which was compared with that of previously reported complex 2 ([PtI(2,9-dimethyl-1,10-phenanthroline)(1-methyl-cytosine)]I) and oxaliplatin and cisplatin as reference compounds. The cytotoxicity data were correlated with the cellular uptake and the DNA platination levels. Finally, the reactivity of 1 towards guanosine 5'-monophosphate (5'-GMP) and glutathione was investigated to provide insights into its mechanism of action.
Sigma-Aldrich
Oxaliplatin, powder
Supelco
Oxaliplatin, Pharmaceutical Secondary Standard; Certified Reference Material
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
USP
Oxaliplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%