Merck
  • Effect of ursodeoxycholic acid treatment on the altered progesterone and bile acid homeostasis in the mother-placenta-foetus trio during cholestasis of pregnancy.
Effect of ursodeoxycholic acid treatment on the altered progesterone and bile acid homeostasis in the mother-placenta-foetus trio during cholestasis of pregnancy.
British journal of clinical pharmacology (2014-08-08)
Maria C Estiú, Maria J Monte, Laura Rivas, Maria Moirón, Laura Gomez-Rodriguez, Tomas Rodriguez-Bravo, Jose J G Marin, Rocio I R Macias
ABSTRACT
Intrahepatic cholestasis of pregnancy (ICP) is characterized by pruritus and elevated bile acid concentrations in maternal serum. This is accompanied by an enhanced risk of intra-uterine and perinatal complications. High concentrations of sulphated progesterone metabolites (PMS) have been suggested to be involved in the multifactorial aetiopathogenesis of ICP. The aim of this study was to investigate further the mechanism accounting for the beneficial effect of oral administration of ursodeoxycholic acid (UDCA), which is the standard treatment, regarding bile acid and PMS homeostasis in the mother-placenta-foetus trio. Using HPLC-MS/MS bile acids and PMS were determined in maternal and foetal serum and placenta. The expression of ABC proteins in placenta was determined by real time quantitative PCR (RT-QPCR) and immunofluorescence. In ICP, markedly increased concentrations of bile acids (tauroconjugates > glycoconjugates > unconjugated), progesterone and PMS in placenta and maternal serum were accompanied by enhanced concentrations in foetal serum of bile acids, but not of PMS. UDCA treatment reduced bile acid accumulation in the mother-placenta-foetus trio, but had no significant effect on progesterone and PMS concentrations. ABCG2 mRNA abundance was increased in placentas from ICP patients vs. controls and remained stable following UDCA treatment, despite an apparent further increase in ABCG2. UDCA administration partially reduces ICP-induced bile acid accumulation in mothers and foetuses despite the lack of effect on concentrations of progesterone and PMS in maternal serum. Up-regulation of placental ABCG2 may play an important role in protecting the foetus from high concentrations of bile acids and PMS during ICP.
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
6-Maleimidohexanoic acid, ≥98.0% (HPLC)
Sigma-Aldrich
5β-Pregnan-3α-ol-20-one
Sigma-Aldrich
5α-Pregnan-3α-ol-20-one, solid
Sigma-Aldrich
Lithocholic acid, ≥95%
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Chenodeoxycholic acid
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Lithocholic acid, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)
Estriol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)