Merck
  • Coincidence detection in a neural correlate of classical conditioning is initiated by bidirectional 3-phosphoinositide-dependent kinase-1 signalling and modulated by adenosine receptors.
Coincidence detection in a neural correlate of classical conditioning is initiated by bidirectional 3-phosphoinositide-dependent kinase-1 signalling and modulated by adenosine receptors.
The Journal of physiology (2015-02-03)
Joyce Keifer, Zhaoqing Zheng
ABSTRACT
How the neural substrates for detection of paired stimuli are distinct from unpaired stimuli is poorly understood and a fundamental question for understanding the signalling mechanisms for coincidence detection during associative learning. To address this question, we used a neural correlate of eyeblink classical conditioning in an isolated brainstem from the turtle, in which the cranial nerves are directly stimulated in place of using a tone or airpuff. A bidirectional response is activated in <5 min of training, in which phosphorylated 3-phosphoinositide-dependent kinase-1 (p-PDK1) is increased in response to paired and decreased in response to unpaired nerve stimulation and is mediated by the opposing actions of neurotrophin receptors TrkB and p75(NTR) . Surprisingly, blockade of adenosine 2A (A2A ) receptors inhibits both of these responses. Pairing also induces substantially increased surface expression of TrkB that is inhibited by Src family tyrosine kinase and A2A receptor antagonists. Finally, the acquisition of conditioning is blocked by a PDK1 inhibitor. The unique action of A2A receptors to function directly as G proteins and in receptor transactivation to control distinct TrkB and p75(NTR) signalling pathways allows for convergent activation of PDK1 and protein kinase A during paired stimulation to initiate classical conditioning.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SAFC
Glycine
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%