Merck
Development of antileishmanial lipid nanocomplexes.
Biochimie (2014-06-22)
T T H Pham, C Gueutin, M Cheron, S Abreu, P Chaminade, P M Loiseau, G Barratt
ABSTRACT
Visceral leishmaniasis is a life-threatening disease that affects nearly a million people every year. The emergence of Leishmania strains resistant to existing drugs complicates its treatment. The purpose of this study was to develop a new lipid formulation based on nanocochleates combining two active drugs: Amphotericin B (AmB) and Miltefosine (HePC). Nanocochleates composed of dioleoylphosphatidylserine (DOPS) and Cholesterol (Cho) and Ca(2+), in which HePC and AmB were incorporated, were prepared. Properties such as particle size, zeta potential, drug payload, in-vitro drug release and storage stability were investigated. Moreover, in-vitro stability in gastrointestinal fluid was performed in view of an oral administration. AmB-HePC-loaded nanocochleates with a mean particle size of 250 ± 2 nm were obtained. The particles displayed a narrow size distribution and a drug payload of 29.9 ± 0.5 mg/g for AmB, and 14.0 ± 0.9 mg/g for HePC. Drug release occurred preferentially in intestinal medium containing bile salts. Therefore, AmB-HePC-loaded nanocochleates could be a promising oral delivery system for the treatment of visceral leishmaniasis.
Product Description
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Maleic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Maleic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Maleic acid, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)