Merck
Sepsis attenuates the anabolic response to skeletal muscle contraction.
Shock (Augusta, Ga.) (2014-11-26)
Jennifer L Steiner, Charles H Lang
ABSTRACT
Electrically stimulated muscle contraction is a potential clinical therapy to treat sepsis-induced myopathy; however, whether sepsis alters contraction-induced anabolic signaling is unknown. Polymicrobial peritonitis was produced by cecal ligation and puncture (CLP) in male C57BL/6 mice and time-matched, pair-fed controls (CON). At ∼24 h post-CLP, the right hindlimb was electrically stimulated via the sciatic nerve to evoke maximal muscle contractions, and the gastrocnemius was collected 2 h later. Protein synthesis was increased by muscle contraction in CON mice. Sepsis suppressed the rate of synthesis in both the nonstimulated (31%) and stimulated (57%) muscle versus CON. Contraction of muscle in CON mice increased the phosphorylation of mTORC1 (mammalian target of rapamycin [mTOR] complex 1) substrates S6K1 (70-kd ribosomal protein S6 kinase 1) Thr (8-fold), S6K1 ThrSer (7-fold) and 4E-BP1 Ser (11-fold). Sepsis blunted the contraction-induced phosphorylation of S6K1 Thr (67%), S6K1 ThrSer (46%), and 4E-BP1 Ser (85%). Conversely, sepsis did not appear to modulate protein elongation as eEF2 Thr phosphorylation was decreased similarly by muscle contraction in both groups. Mitogen-activated protein kinase signaling was discordant following contraction in septic muscle; phosphorylation of extracellular signal-regulated kinase ThrTyr and p38 ThrTyr was increased similarly in both CON and CLP mice, while sepsis prevented the contraction-induced phosphorylation of JNK ThrTyr and c-JUN Ser. The expression of interleukin 6 and tumor necrosis factor α (TNF-α) mRNA in muscle was increased by sepsis, and contraction increased TNF-α to a greater extent in muscle from septic than CON mice. Injection of the mTOR inhibitor Torin2 in separate mice confirmed that contraction-induced increases in S6K1 and 4E-BP1 were mTOR mediated. These findings demonstrate that resistance to contraction-induced anabolic signaling occurs during sepsis and is predominantly mTORC1-dependent.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium Fluoride Solution
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, suitable for HPLC, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethanol
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC