Merck
  • Protective effect of calpain inhibitor N-acetyl-L-leucyl-L-leucyl-L-norleucinal on acute alcohol consumption related cardiomyopathy.
Protective effect of calpain inhibitor N-acetyl-L-leucyl-L-leucyl-L-norleucinal on acute alcohol consumption related cardiomyopathy.
Molecular biology reports (2014-07-06)
Kazim Kartkaya, Güngör Kanbak, Ayşegül Oğlakçı, Dilek Burukoğlu, Mehmet Caner Ozer
ABSTRACT
Excessive alcohol consumption and alcoholism cause medical problems with high mortality and morbidity rates. In this study we aimed to decrease the alcohol related tissue damage by inhibiting calpain activation which plays an important role in apoptosis and necrosis, in rats with cardiomyopathy induced by acute alcohol consumption. Male Sprague-Dawley rats were separated into four groups (control, vehicle, alcohol and alcohol + inhibitor) with 10 rats in each. Control group received isocaloric maltose while vehicle group received isocaloric maltose with DMSO, and alcohol group received 8 g/kg absolute ethanol by gavage. Inhibitor group received 20 mg/kg calpain inhibitor 1 intraperitonally prior to alcohol administration. Calpain activities, cathepsin L levels and cytochrome c release rates were significantly increased in alcohol group compared to control group (p < 0.05). Serum CK MB and BNP levels of alcohol group were excessively increased compared to control group (respectively p < 0.001 and p < 0.01). Serum BNP levels of alcohol + inhibitor group were significantly (p < 0.05) decreased compared to alcohol group. In addition to these, histological evaluation of light microscope images and the results of DNA fragmentation and immunohistochemical caspase-3 activity results showed significant improvement of these parameters in alcohol + inhibitor group compared to alcohol group. Results of our biochemical and histological evaluation results revealed that the calpain inhibitor N-acetyl-leu-leu-norleucinal may have an ameliorating effect on acute alcohol consumption related cardiac tissue damage due to its effects on cell death pathways.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture