  • Search Results
  • Quantitative proteomics to characterize specific histone H2A proteolysis in chronic lymphocytic leukemia and the myeloid THP-1 cell line.
Quantitative proteomics to characterize specific histone H2A proteolysis in chronic lymphocytic leukemia and the myeloid THP-1 cell line.
International journal of molecular sciences (2014-05-30)
Pieter Glibert, Liesbeth Vossaert, Katleen Van Steendam, Stijn Lambrecht, Filip Van Nieuwerburgh, Fritz Offner, Thomas Kipps, Maarten Dhaenens, Dieter Deforce
ABSTRACT
Proteome studies on hematological malignancies contribute to the understanding of the disease mechanism and to the identification of new biomarker candidates. With the isobaric tag for relative and absolute quantitation (iTRAQ) method we analyzed the protein expression between B-cells of healthy people and chronic lymphocytic leukemia (CLL) B-cells. CLL is the most common lymphoid cancer of the blood and is characterized by a variable clinical course. By comparing samples of patients with an aggressive vs. indolent disease, we identified a limited list of differentially regulated proteins. The enhanced sensitivity attributed to the iTRAQ labels led to the discovery of a previously reported but still not clarified proteolytic product of histone H2A (cH2A) which we further investigated in light of the suggested functional properties of this modification. In the exploratory proteome study the Histone H2A peptide was up-regulated in CLL samples but a more specific and sensitive screening of a larger patient cohort indicated that cH2A is of myeloid origin. Our subsequent quantitative analysis led to a more profound characterization of the clipping in acute monocytic leukemia THP-1 cells subjected to induced differentiation.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
CAPS, ≥99%
Sigma-Aldrich
CAPS, BioXtra, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
CAPS, BioUltra, ≥99.0% (TLC)
Sigma-Aldrich
Tributylphosphine, ≥93.5% (Tri-N-butylphosphine, GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Tributylphosphine, mixture of isomers, 97%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%