Merck
  • HDAC inhibitors target HDAC5, upregulate microRNA-125a-5p, and induce apoptosis in breast cancer cells.
HDAC inhibitors target HDAC5, upregulate microRNA-125a-5p, and induce apoptosis in breast cancer cells.
Molecular therapy : the journal of the American Society of Gene Therapy (2014-12-23)
Tsung-Hua Hsieh, Chia-Yi Hsu, Cheng-Fang Tsai, Cheng-Yu Long, Chin-Hu Wu, Deng-Chyang Wu, Jau-Nan Lee, Wei-Chun Chang, Eing-Mei Tsai
ABSTRACT
Histone deacetylase inhibitors (HDACi) are novel clinical anticancer drugs that inhibit HDAC gene expression and induce cell apoptosis in human cancers. Nevertheless, the detailed mechanism or the downstream HDAC targets by which HDACi mediates apoptosis in human breast cancer cells remains unclear. Here, we show that HDACi reduce tumorigenesis and induce intrinsic apoptosis of human breast cancer cells through the microRNA miR-125a-5p in vivo and in vitro. Intrinsic apoptosis was activated by the caspase 9/3 signaling pathway. In addition, HDACi mediated the expression of miR-125a-5p by activating RUNX3/p300/HDAC5 complex. Subsequently, miR-125a-5p silenced HDAC5 post-transcriptionally in the cells treated with HDACi. Thus, a regulatory loop may exist in human breast cancer cells involving miR-125a-5p and HDAC5 that is controlled by RUNX3 signaling. Silencing of miR-125a-5p and RUNX3 inhibited cancer progression and activated apoptosis, but silencing of HDAC5 had a converse effect. In conclusion, we demonstrate a possible new mechanism by which HDACi influence tumorigenesis and apoptosis via downregulation of miR-125a-5p expression. This study provides clinical implications in cancer chemotherapy using HDACi.
Product Description
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propylpentanoic acid
Sigma-Aldrich
Nα-Acetyl-L-lysine
Sigma-Aldrich
M344, ≥98% (HPLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Valproic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Supelco
Sodium butyrate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Apicidin, ≥98% (HPLC), from microbial