Merck
  • Ultra-high performance liquid chromatography tandem mass spectrometric method for the determination of tamoxifen, N-desmethyltamoxifen, 4-hydroxytamoxifen and endoxifen in dried blood spots--development, validation and clinical application during breast cancer adjuvant therapy.
Ultra-high performance liquid chromatography tandem mass spectrometric method for the determination of tamoxifen, N-desmethyltamoxifen, 4-hydroxytamoxifen and endoxifen in dried blood spots--development, validation and clinical application during breast cancer adjuvant therapy.
Talanta (2014-12-06)
Marina Venzon Antunes, Suziane Raymundo, Vanessa de Oliveira, Dilana Elisabeth Staudt, Gustavo Gössling, Giovana Piva Peteffi, Jorge Villanova Biazús, José Antônio Cavalheiro, Marie Tre-Hardy, Arnaud Capron, Vincent Haufroid, Pierre Wallemacq, Gilberto Schwartsmann, Rafael Linden
ABSTRACT
A LC-MSMS method for the simultaneous determination of tamoxifen, N-desmethyltamoxifen, 4-hydroxytamoxifen and endoxifen in dried blood spots samples was developed and validated. The method employs an ultrasound-assisted liquid extraction and a reversed phase separation in an Acquity(®) C18 column (150×2.1 mm, 1.7 µm). Mobile phase was a mixture of formic acid 0.1% (v/v) pH 2.7 and acetonitrile (gradient from 60:40 to 50:50, v/v). Total analytical run time was 8 min. Precision assays showed CV % lower than 10.75% and accuracy in the range 94.5 to 110.3%. Mean analytes recoveries from DBS ranged from 40% to 92%. The method was successfully applied to 91 paired clinical DBS and plasma samples. Dried blood spots concentrations were highly correlated to plasma, with rs>0.83 (P<0.01). Median estimated plasma concentrations after hematocrit and partition factor adjustment were: TAM 123.3 ng mL(-1); NDT 267.9 ng mL(-1), EDF 10.0 ng mL(-1) and HTF 1.3 ng mL(-1,) representing in average 98 to 104% of the actually measured concentrations. The DBS method was able to identify 96% of patients with plasma EDF concentrations below the clinical threshold related to better prognosis (5.9 ng mL(-1)). The procedure has adequate analytical performance and can be an efficient tool to optimize adjuvant breast cancer treatment, especially in resource limited settings.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), analytical standard
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
USP
1-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material