Merck
  • Biodegradable salicylate-based poly(anhydride-ester) microspheres for controlled insulin delivery.
Biodegradable salicylate-based poly(anhydride-ester) microspheres for controlled insulin delivery.
Journal of biomedical materials research. Part A (2013-09-13)
Roberto Delgado-Rivera, Roselin Rosario-Meléndez, Weiling Yu, Kathryn E Uhrich
ABSTRACT
Salicylate-based poly(anhydride-esters) (PAEs) chemically incorporate salicylic acid (SA) into the polymer backbone, which is then delivered in a controlled manner upon polymer hydrolysis. In this work, a salicylate-based PAE is a carrier to encapsulate and deliver insulin. Polymer microspheres were formulated using a water/oil/water double-emulsion solvent evaporation technique. The microspheres obtained had a smooth surface, high protein encapsulation efficiency, and relatively low emulsifier content. Insulin was released in vitro for 15 days, with no signs of aggregation or unfolding of the secondary structure. The released insulin also retained bioactivity in vitro. Concurrently, SA was released from the microspheres with polymer degradation and anti-inflammatory activity was observed. Based upon these results, the formulated microspheres enable simultaneous delivery of insulin and SA, both retaining bioactivity following processing.
Product Description
SAFC
L-Glutamine
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source