Merck
  • Isolation and characterization of Bacillus subtilis CH16 strain from chicken gastrointestinal tracts for use as a feed supplement to promote weight gain in broilers.
Isolation and characterization of Bacillus subtilis CH16 strain from chicken gastrointestinal tracts for use as a feed supplement to promote weight gain in broilers.
Letters in applied microbiology (2015-03-11)
A T V Nguyen, D V Nguyen, M T Tran, L T Nguyen, A H Nguyen, T-N Phan
ABSTRACT
Spore-forming bacterial strains were isolated from chicken gastrointestinal tracts to develop a heat-stable feed supplement that promotes weight gain in broilers. Seven Bacillus strains having more than 90% sporulation were screened from the isolates and identified to be closely related with Bacillus subtilis and Bacillus licheniformis. Of the seven strains, B. subtilis CH16 was selected to develop a feed supplement for broilers, because it formed 100% heat-stable spores, grew rapidly at 42°C and quickly formed a biofilm. In large-scale trials in broilers (n ≥ 1150 per group), the group fed CH16 (3 × 10(6) CFU g(-1) pellet) showed higher average daily gain (ADG = 61·16) and lower food conversion ratio (FCR = 1·696) than did the group fed B. licheniformis CH22 (ADG = 57·10 and FCR = 1·792), the group fed B. subtilis HU58 (ADG = 51·90 and FCR = 1·868), BioPlus group (ADG = 59·32 and FCR = 1·807) and the control group (ADG = 56·02 and FCR = 1·880). In conclusion, CH16 spores significantly increased ADG by 9·17% and reduced FCR by 9·79% in broilers. The result supports the use of B. subtilis CH16 of chicken intestinal origin as a feed supplement that promote weight gain in broilers. Significance and impact of the study: This study reports screening of Bacillus strains isolated from chicken gastrointestinal tracts for development of a feed supplement that promote weight gain in broilers. Of the seven Bacillus isolates with high sporulation efficiency (≥90%), Bacillus subtilis CH16 strain showed the best growth and biofilm formation at body temperature of broilers (42°C). In large-scale trials in broilers (n ≥ 1150 per group), CH16 spores induced a 9·17% increase in daily weight gain (ADG) and a 9·79% reduction in FCR while the commercial BioPlus(®) YC induced only a 5·89% increase in ADG and a 3·88% reduction in FCR.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M