Merck
  • Quantitative determination of dopamine in single rat pheochromocytoma cells by microchip electrophoresis with only one high-voltage power supply.
Quantitative determination of dopamine in single rat pheochromocytoma cells by microchip electrophoresis with only one high-voltage power supply.
Journal of separation science (2015-04-22)
Cuicui Sha, Yuejuan Fan, Jieke Cheng, Han Cheng
ABSTRACT
We developed a method for the direct identification of dopamine in single cultured rat pheochromocytoma cells by capillary electrophoresis using an end-channel carbon fiber nanoelectrode amperometric detector. The operation mode was designed to achieve single-cell injection and lysis in microfluidic chip electrophoresis with only one high-voltage power supply. The separation and detection conditions were optimized. Four catecholamines were baseline-separated and determined with this system, and the cell density and liquid height of the reservoirs were accommodated for single cell loading, docking and analysis. The microchip capillary electrophoresis system was successfully applied to determine dopamine in single cultured rat pheochromocytoma cells.
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%