  • Search Results
  • Activity-guided chemo toxic profiling of Cassia occidentalis (CO) seeds: detection of toxic compounds in body fluids of CO-exposed patients and experimental rats.
Activity-guided chemo toxic profiling of Cassia occidentalis (CO) seeds: detection of toxic compounds in body fluids of CO-exposed patients and experimental rats.
Chemical research in toxicology (2015-04-29)
Gati Krushna Panigrahi, Ratnasekhar Ch, Mohana K R Mudiam, Vipin M Vashishtha, S Raisuddin, Mukul Das
ABSTRACT
Our prior studies have shown an association between the deaths of children and consumption of Cassia occidentalis (CO) seeds. However, the chemicals responsible for the CO poisoning are not known. Therefore, the present study was designed to identify the key moieties in CO seeds and their cytotoxicity in rat primary hepatocytes and HepG2 cells. Activity-guided sequential extraction and fractionation of the seeds followed by GC-MS analysis identified the toxic compounds in the CO seeds. These identified compounds were subsequently detected and quantified in blood and urine samples from CO-exposed rats and CO poisoning human study cases. GC-MS analysis of different fractions of methanol extracts of CO seeds revealed the presence of five anthraquinones (AQs), viz. physcion, emodin, rhein, aloe-emodin, and chrysophanol. Interestingly, these AQs were detected in serum and urine samples from the study cases and CO-exposed rats. Cytotoxicity analysis of the above AQs in rat primary hepatocytes and HepG2 cells revealed that rhein is the most toxic moiety, followed by emodin, aloe-emodin, physcion, and chrysophanol. These studies indicate that AQ aglycones are responsible for producing toxicity, which may be associated with symptoms of hepatomyoencephalopathy in CO poisoning cases.
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Emodin, from Frangula bark, ≥90% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Physcion, ≥98.0% (TLC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
1,8-Dihydroxy-3-methylanthraquinone, 98%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%