Merck
  • A Conformational Shift in the Dissociated Cholera Toxin A1 Subunit Prevents Reassembly of the Cholera Holotoxin.
A Conformational Shift in the Dissociated Cholera Toxin A1 Subunit Prevents Reassembly of the Cholera Holotoxin.
Toxins (2015-08-13)
Michael Taylor, David Curtis, Ken Teter
ABSTRACT
Cholera toxin (CT) consists of a catalytic A1 subunit, an A2 linker, and a homopentameric cell-binding B subunit. The intact holotoxin moves by vesicle carriers from the cell surface to the endoplasmic reticulum (ER) where CTA1 is released from the rest of the toxin. The dissociated CTA1 subunit then shifts to an unfolded conformation, which triggers its export to the cytosol by a process involving the quality control system of ER-associated degradation (ERAD). We hypothesized that the unfolding of dissociated CTA1 would prevent its non-productive reassociation with CTA2/CTB5. To test this prediction, we monitored the real-time reassociation of CTA1 with CTA2/CTB5 by surface plasmon resonance. Folded but not disordered CTA1 could interact with CTA2/CTB5 to form a stable, functional holotoxin. Our data, thus, identified another role for the intrinsic instability of the isolated CTA1 polypeptide in host-toxin interactions: in addition to activating the ERAD translocation mechanism, the spontaneous unfolding of free CTA1 at 37 °C prevents the non-productive reassembly of a CT holotoxin in the ER.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
CTB, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis