Merck
  • Insights into Protein Structure and Dynamics by Ultraviolet and Visible Resonance Raman Spectroscopy.
Insights into Protein Structure and Dynamics by Ultraviolet and Visible Resonance Raman Spectroscopy.
Biochemistry (2015-07-30)
Ignacio López-Peña, Brian S Leigh, Diana E Schlamadinger, Judy E Kim
ABSTRACT
Raman spectroscopy is a form of vibrational spectroscopy based on inelastic scattering of light. In resonance Raman spectroscopy, the wavelength of the incident light falls within an absorption band of a chromophore, and this overlap of excitation and absorption energy greatly enhances the Raman scattering efficiency of the absorbing species. The ability to probe vibrational spectra of select chromophores within a complex mixture of molecules makes resonance Raman spectroscopy an excellent tool for studies of biomolecules. In this Current Topic, we discuss the type of molecular insights obtained from steady-state and time-resolved resonance Raman studies of a prototypical photoactive protein, rhodopsin. We also review recent efforts in ultraviolet resonance Raman investigations of soluble and membrane-associated biomolecules, including integral membrane proteins and antimicrobial peptides. These examples illustrate that resonance Raman is a sensitive, selective, and practical method for studying the structures of biological molecules, and the molecular bonding, geometry, and environments of protein cofactors, the backbone, and side chains.
Sigma-Aldrich
Chlorophyll a, from spinach
Sigma-Aldrich
Chlorophyll a, from Anacystis nidulans algae
Sigma-Aldrich
Melittin from honey bee venom, ≥65% (HPLC)
Sigma-Aldrich
Melittin, ≥97% (HPLC), synthetic
Sigma-Aldrich
Melittin from honey bee venom, ≥85% (HPLC)