Merck
  • Two new triterpenoid saponins from the aerial parts of Anemone taipaiensis.
Two new triterpenoid saponins from the aerial parts of Anemone taipaiensis.
Journal of Asian natural products research (2015-05-30)
Hui Li, Xiao-Yang Wang, Xia-Yin Wang, Dong Hua, Yang Liu, Hai-Feng Tang
ABSTRACT
Phytochemical study on the aerial parts of Anemone taipaiensis for the first time led to the isolation of two new oleanane-type triterpenoid saponins 1 and 2, together with four known saponins (3-6). Their structures were elucidated by extensive spectroscopic analysis and chemical evidences. Saponins 2-4 exhibited cytotoxicity against human glioblastoma U251MG cell line with IC50 values ranging from 1.56 to 80.62 μM.
Sigma-Aldrich
Pyridine-d5, ≥99.5 atom % D
Sigma-Aldrich
Pyridine-d5, "100%", ≥99.96 atom % D
Sigma-Aldrich
Pyridine-d5, ≥99.5 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)