Merck
  • shRNA-Mediated Silencing of Y-Box Binding Protein-1 (YB-1) Suppresses Growth of Neuroblastoma Cell SH-SY5Y In Vitro and In Vivo.
shRNA-Mediated Silencing of Y-Box Binding Protein-1 (YB-1) Suppresses Growth of Neuroblastoma Cell SH-SY5Y In Vitro and In Vivo.
PloS one (2015-05-21)
Hong Wang, Ruowen Sun, Min Gu, Shuang Li, Bin Zhang, Zuofei Chi, Liangchun Hao
ABSTRACT
Y-box binding protein-1 (YB-1), a member of cold-shock protein superfamily, has been demonstrated to be associated with tumor malignancy, and is proposed as a prognostic marker in multiple carcinomas. However, the role of YB-1 in neuroblastoma has not been well studied. To investigate the functional role of YB-1 in neuroblastoma, we established a YB-1-silenced neuroblastoma cell strain by inhibiting YB-1 expression using a shRNA knockdown approach. YB-1-silenced neuroblastoma SH-SY5Y cells exhibited a pronounced reduction in cell proliferation and an increased rate of apoptosis in vitro and in vivo xenograft tumor model. At molecular level, YB-1 silencing resulted in downregulation of Cyclin A, Cyclin D1 and Bcl-2, as well as upregulated levels of Bax, cleaved caspase-3 and cleaved PARP-1. We further demonstrated that YB-1 transcriptionally regulated Cyclin D1 expression by chromatin-immunoprecipitation and luciferase reporter assays. In addition, xenograft tumors derived from neuroblastoma SH-SY5Y cell line were treated with YB-1 shRNA plasmids by intra-tumor injection, and YB-1 targeting effectively inhibited tumor growth and induced cell death. In summary, our findings suggest that YB-1 plays a critical role in neuroblastoma development, and it may serve as a potential target for neuroblastoma therapy.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
EZ-Zyme Chromatin Prep Kit, Contains proprietary reagents optimized for the enzymatic shearing of chromatin from mammalian cells at higher resolution than sonication for use in chromatin immunoprecipitation (ChIP) assays.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution