Merck
  • Stimuli-Triggered Activity of Nanoreactors by Biomimetic Engineering Polymer Membranes.
Stimuli-Triggered Activity of Nanoreactors by Biomimetic Engineering Polymer Membranes.
Nano letters (2015-10-03)
Tomaž Einfalt, Roland Goers, Ionel Adrian Dinu, Adrian Najer, Mariana Spulber, Ozana Onaca-Fischer, Cornelia G Palivan
ABSTRACT
The development of advanced stimuli-responsive systems for medicine, catalysis, or technology requires compartmentalized reaction spaces with triggered activity. Only very few stimuli-responsive systems preserve the compartment architecture, and none allows a triggered activity in situ. We present here a biomimetic strategy to molecular transmembrane transport by engineering synthetic membranes equipped with channel proteins so that they are stimuli-responsive. Nanoreactors with triggered activity were designed by simultaneously encapsulating an enzyme inside polymer compartments, and inserting protein "gates" in the membrane. The outer membrane protein F (OmpF) porin was chemically modified with a pH-responsive molecular cap to serve as "gate" producing pH-driven molecular flow through the membrane and control the in situ enzymatic activity. This strategy provides complex reaction spaces necessary in "smart" medicine and for biomimetic engineering of artificial cells.
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%