Merck
  • Isolation and Identification of a Red Pigment from the Antarctic Bacterium Shewanella frigidimarina.
Isolation and Identification of a Red Pigment from the Antarctic Bacterium Shewanella frigidimarina.
Protein and peptide letters (2015-09-16)
Maria Luisa Martín-Cerezo, Eva García-López, Cristina Cid
ABSTRACT
The present study dealt with the isolation, identification and characterization of pigments from red snow samples of the Quito coastal front glacier (S 62º 27,217', W 059º 45,960') in Greenwich, Archipelago South Shetland, Antarctica, during summer 2013. As a strain of Shewanella was found to be the most common and abundant species with maximum red color production, the pigment -contained in the protein fraction- was isolated and characterized by high performance liquid chromatography (HPLC), two-dimensional fluorescence Difference Gel Electrophoresis (2-D DIGE) and matrix- assisted laser desorption/ionization-time of flight mass spectrometry (MALDI/TOF/TOF). The identified pigment is a cytochrome c3 with apparent molecular weight of 10 kDa and apparent pI around 4.5. The maximum pigment concentration was produced at warm temperatures, 28ºC, and with increasing exposure time to UV radiation. Here we demonstrate that the synthesis of cytochrome c3 by the Antarctic bacterium is due to thermal adaptation and/or adaptation to radiation. Further, pigments such as cytochrome c3 enable this bacterial species to use an anaerobic and ferric metabolism. In addition, this study draws attention to the relevance of adaptation investigations; to the study of in vivo monitoring of environmental warming and UV radiation due to global warming; and to the study of the potential habitability of other worlds in the Solar System and beyond.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%