Merck
  • Solvent modulation strategy for superior antibody monomer/aggregate separation in cation exchange chromatography.
Solvent modulation strategy for superior antibody monomer/aggregate separation in cation exchange chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-11-02)
Simon Kluters, Christian Frech, Thomas von Hirschheydt, Andreas Schaubmar, Sebastian Neumann
ABSTRACT
Cation exchange chromatography (CEX) is an integral part of many downstream processes for monoclonal antibodies (mAbs). However, in some cases CEX methods with standard mobile phase conditions do not lead to a sufficient removal of soluble antibody aggregates. The addition of neutral polymers such as polyethylene glycol (PEG) to the mobile phase can improve the separation of proteins in IEC remarkably. The applicability of this solvent modulation technique is limited by protein precipitation at higher PEG concentrations. To overcome this limitation solubility enhancers like polyols and amino acids can be added to the mobile phase. These additives are known to inhibit PEG-induced protein precipitation in solution. This new solvent modulation strategy was tested with three different mAbs on two different CEX resins in the presence of PEG in combination with various solubility enhancers. In order to assess the general applicability of this method, mAbs were selected that show major differences with respect to their sensitivity to PEG-induced precipitation and monomer/aggregate resolution performance that is achieved by CEX under standard conditions. For all three mAbs precipitation could be prevented without elimination of the positive PEG-effect. The addition of solubility enhancers gives access to improved separation at elevated PEG concentrations and high protein loadings without running into precipitation issues. Our data indicate that this method is generically applicable and leads to a superior antibody monomer/aggregate separation.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
L-Proline
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Glycine
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)