Merck
  • Epigenetic Readers of Lysine Acetylation Regulate Cocaine-Induced Plasticity.
Epigenetic Readers of Lysine Acetylation Regulate Cocaine-Induced Plasticity.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-11-13)
Gregory C Sartor, Samuel K Powell, Shaun P Brothers, Claes Wahlestedt
ABSTRACT
Epigenetic processes that regulate histone acetylation play an essential role in behavioral and molecular responses to cocaine. To date, however, only a small fraction of the mechanisms involved in the addiction-associated acetylome have been investigated. Members of the bromodomain and extraterminal (BET) family of epigenetic "reader" proteins (BRD2, BRD3, BRD4, and BRDT) bind acetylated histones and serve as a scaffold for the recruitment of macromolecular complexes to modify chromatin accessibility and transcriptional activity. The role of BET proteins in cocaine-induced plasticity, however, remains elusive. Here, we used behavioral, pharmacological, and molecular techniques to examine the involvement of BET bromodomains in cocaine reward. Of the BET proteins, BRD4, but not BRD2 or BRD3, was significantly elevated in the nucleus accumbens (NAc) of mice and rats following repeated cocaine injections and self-administration. Systemic and intra-accumbal inhibition of BRD4 with the BET inhibitor, JQ1, attenuated the rewarding effects of cocaine in a conditioned place preference procedure but did not affect conditioned place aversion, nor did JQ1 alone induce conditioned aversion or preference. Investigating the underlying mechanisms, we found that repeated cocaine injections enhanced the binding of BRD4, but not BRD3, to the promoter region of Bdnf in the NAc, whereas systemic injection of JQ1 attenuated cocaine-induced expression of Bdnf in the NAc. JQ1 and siRNA-mediated knockdown of BRD4 in vitro also reduced expression of Bdnf. These findings indicate that disrupting the interaction between BET proteins and their acetylated lysine substrates may provide a new therapeutic avenue for the treatment of drug addiction. Proteins involved in the "readout" of lysine acetylation marks, referred to as BET bromodomain proteins (including BRD2, BRD3, BRD4, and BRDT), have been shown to be key regulators of chromatin dynamics and disease, and BET inhibitors are currently being studied in several clinical trials. However, their role in addiction-related phenomena remains unknown. In the current studies, we revealed that BRD4 is elevated in the nucleus accumbens and recruited to promoter regions of addiction-related genes following repeated cocaine administration, and that inhibition of BRD4 attenuates transcriptional and behavioral responses to cocaine. Together, these studies reveal that BET inhibitors may have therapeutic utility in the treatment of cocaine addiction.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dner
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptprj
Sigma-Aldrich
MISSION® esiRNA, targeting human DNER
Sigma-Aldrich
MISSION® esiRNA, targeting human PTPRJ
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%