  • Search Results
  • Exploring the possibilities of capacitively coupled contactless conductivity detection in combination with liquid chromatography for the analysis of polar compounds using aminoglycosides as test case.
Exploring the possibilities of capacitively coupled contactless conductivity detection in combination with liquid chromatography for the analysis of polar compounds using aminoglycosides as test case.
Journal of pharmaceutical and biomedical analysis (2015-01-01)
Péter Jankovics, Shruti Chopra, Mohamed N El-Attug, Deirdre Cabooter, Kris Wolfs, Béla Noszál, Ann Van Schepdael, Erwin Adams
ABSTRACT
The analysis of highly polar (often charged) compounds which lack a strong UV absorbing chromophore is really challenging. Despite the numerous analytical methods published, the demand for a simple, robust and cheap technique for their analysis still persists. Here, reversed phase (RP) liquid chromatography (LC) with capacitively coupled contactless conductivity detection (C(4)D) was explored for the first time as a possible method for separation and detection of various aminoglycoside (AMG) antibiotics which were taken as typical test compounds: tobramycin (TOB), spectinomycin, streptomycin, amikacin, kanamycin A and kanamycin B. C(4)D was performed using a commercially available as well as a laboratory made cell. As ion-pairing reagents (IPR) four perfluorinated carboxylic acids were used: pentafluoropropionic acid, heptafluorobutyric acid, nonafluoropentanoic acid (NFPA) and pentadecafluorooctanoic acid (PDFOA). 0.125 mM NFPA-acetonitrile (ACN) (90:10) or 0.125 mM PDFOA-ACN (70:30) as mobile phases were suitable to detect TOB with reasonable retention times. However, NFPA was preferred for practical reasons. Its applicable concentration range in the mobile phase was strongly restricted by loss of chromatographic performance at lower levels and excessive background conductivity at higher levels. Overall repeatability and robustness of the method were rather poor which was explained by the relatively low IPR levels. Selectivity between the tested AMGs was mainly influenced by the number of protonated amino groups per molecule making it impossible to separate compounds of equal net charges. Problems encountered with gradient elution, hydrophilic interaction liquid chromatography (HILIC) and separation at high pH without IPRs are also discussed.
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Perfluoropentanoic acid, 97%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
DL-Valine, ≥97%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Valine, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Pentafluoropropionic acid, 97%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
2,2,3,3,3-Pentafluoropropyl acrylate, 98%, contains 100 ppm 4-tert-butylcatechol as inhibitor
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)