Merck
  • The p53-inducible gene 3 involved in flavonoid-induced cytotoxicity through the reactive oxygen species-mediated mitochondrial apoptotic pathway in human hepatoma cells.
The p53-inducible gene 3 involved in flavonoid-induced cytotoxicity through the reactive oxygen species-mediated mitochondrial apoptotic pathway in human hepatoma cells.
Food & function (2015-03-31)
Qiang Zhang, Guangdong Cheng, Hongbin Qiu, Liling Zhu, Zhongjuan Ren, Wei Zhao, Tao Zhang, Lei Liu
ABSTRACT
Flavonoids have been reported to exhibit prooxidant cytotoxicity against cancer cells, but the underlying mechanism is still poorly understood. Here we investigated the potential mechanism that p53-inducible gene 3 (PIG3), a NADPH:quinone oxidoreductase, mediated the prooxidant cytotoxicity of flavonoids on human hepatoma HepG2 cells. The results showed that flavonoids (apigenin, luteolin, kaempferol, and quercetin) inhibited the growth of HepG2 cells in a dosage- and time-dependent manner, and induced the morphological changes characteristic of apoptosis in HepG2 cells. We also found that expression of PIG3 was increased markedly in HepG2 cells treated with flavonoids at both mRNA and protein levels, which was accompanied by increased intracellular ROS production and a decreased mitochondrial membrane potential (ΔΨm). All these effects were largely reversed through knockdown of the PIG3 gene in HepG2 cells. Western blotting indicated that flavonoids increased cytochrome c release, upregulated the ratio of Bax/Bcl-2, and activated the caspases-9 and -3. Moreover, knockdown of PIG3 could reverse the changes of these apoptotic-related proteins. These results suggest that PIG3 plays an important role in regulating the prooxidant activity and apoptosis-inducing action of flavonoids on HepG2 cells though the ROS-triggered mitochondrial apoptotic pathway.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)