Merck
  • N-acetylcysteine modulates angiogenesis and vasodilation in stomach such as DNA damage in blood of portal hypertensive rats.
N-acetylcysteine modulates angiogenesis and vasodilation in stomach such as DNA damage in blood of portal hypertensive rats.
World journal of gastroenterology (2015-11-26)
Francielli Licks, Renata Minuzzo Hartmann, Camila Marques, Elizângela Schemitt, Josieli Raskopf Colares, Mariana do Couto Soares, Juliana Reys, Camila Fisher, Juliana da Silva, Norma Possa Marroni
ABSTRACT
To evaluate the antioxidant effect of N-acetylcysteine (NAC) on the stomach of rats with portal hypertension. Twenty-four male Wistar rats weighing ± 250 g were divided into four experimental groups (n = 6 each): Sham-operated (SO), SO + NAC, partial portal vein ligation (PPVL), and PPVL + NAC. Treatment with NAC in a dose of 10 mg/kg (i.p.) diluted in 0.6 mL of saline solution was administered daily for 7 d starting 8 d after the surgery. Animals from the PPVL and SO group received saline solution (0.6 mL) for the same period of time as the PPVL + NAC and SO + NAC group. On the 15(th) day the animals were anesthetized and we evaluated portal pressure by cannulating mesenteric artery. After, we removed the stomach for further analysis. We performed immunohistochemical analysis for endothelial nitric oxide synthase (eNOS), vascular endothelial growth factor (VEGF), and nitrotirosine (NTT) proteins in stomach. We also evaluated eNOS and VEGF by Western blot analysis and assessed DNA damage in blood samples by the comet assay. The portal hypertension group exhibited increases in portal pressure when compared to SO group (29.8 ± 1.8 vs 12.0 ± 0.3 mmHg) (P < 0.001). The same was observed when we compared the eNOS (56.8 ± 3.7 vs 13.46 ± 2.8 pixels) (P < 0.001), VEGF (34.9 ± 4.7 vs 17.46 ± 2.6 pixels) (P < 0.05), and NTT (39.01 ± 4.0 vs 12.77 ± 2.3 pixels) (P < 0.05) expression by immunohistochemistry of the PPVL animals with the SO group. The expression of eNOS (0.39 ± 0.03 vs 0.25 ± 0.03 a.μ) (P < 0.01) and VEGF (0.38 ± 0.04 vs 0.26 ± 0.04 a.μ) (P < 0.01) were also evaluated by Western blot analysis, and we observed an increase of both proteins on PPVL animals. We also evaluated the DNA damage by comet assay, and observed an increase on damage index and damage frequency on those animals. NAC decreased portal pressure values in PPVL + NAC animals (16.46 ± 2 vs 29.8 ± 1.8 mmHg) (P < 0.001) when compared to PPVL. The expression of eNOS (14.60 ± 4.1 vs 56.8 ± 3.7 pixels) (P < 0.001), VEGF (19.53 ± 3.2 vs 34.9 ± 4.7 pixels) (P < 0.05) and NTT (21.84 ± 0.7 vs 39.01 ± 4.0 pixels) (P < 0.05) evaluated by immunohistochemistry were also reduced in PPVL + NAC animals. Also, when evaluated by Western blot eNOS expression (0.32 ± 0.03 vs 0.39 ± 0.03 a.μ) (P < 0.05) and VEGF expression (0.31 ± 0.09 vs 0.38 ± 0.04 a.μ) (P < 0.01). Furthermore, NAC modulated DNA damage in PPVL + NAC animals. In view of these results, we believe NAC is able to protect the stomach from the alterations induced by the PPVL procedure.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis